Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorophenyl isocyanate 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Fluorophenylisocyanate1-Fluoro-3-isocyanatobenzene

L01496-14 L01496-14 L01496-06 L01496-06
AAAL01496-06EA 68.59 USD InStock
CAAAAL01496-06 AAAL01496-06 CAAAAL01496-14 AAAL01496-14
3-Fluorophenyl isocyanate 97+%
3-Fluorophenyl isocyanate


Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 56…58 °C/14 mm
Density: 1.207
Flash Pt: 41 °C(105 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002015
CAS Number: 404-71-7
EINECS: 206-966-1
UN: 3080
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty