Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Heptadecanone 98%

Supplier: Thermo Scientific Chemicals

L01630-06 L01630-14 L01630-14 L01630-06
AAAL01630-06EA 132.99 USD InStock
CAAAAL01630-06 AAAL01630-06 AAAL01630-14 CAAAAL01630-14
9-Heptadecanone 98%
9-Heptadecanone


Formula: C₁₇H₃₄O
MW: 254.46 g/mol
Boiling Pt: 250…253 °C
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00009573
CAS Number: 540-08-9
EINECS: 208-734-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty