Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Fluorophenylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

L01745-MC L01745-ME L01745-04 L01745-MC L01745-ME L01745-04
AAAL01745-MEEA 72.29 USD InStock
AAAL01745-ME CAAAAL01745-MC AAAL01745-04 CAAAAL01745-ME AAAL01745-MC CAAAAL01745-04
α-Fluorophenylacetic acid 98%
α-Fluorophenylacetic acid


Formula: C₈H₇FO₂
MW: 154.14
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004221
CAS Number: 1578-63-8
EINECS: 216-420-4
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty