Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Nitro-1-tetralone 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 7-Nitro-3,4-dihydronaphthalen-1(2H)-one3,4-Dihydro-7-nitronaphthalen-1(2H)-one

L01853-04 L01853-09 L01853-04 L01853-09
AAAL01853-09EA 343.99 USD InStock
AAAL01853-09 CAAAAL01853-09 AAAL01853-04 CAAAAL01853-04
7-Nitro-1-tetralone 97%
7-Nitro-1-tetralone


Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00019661
CAS Number: 40353-34-2
EINECS: 254-887-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty