Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-2,3,5,6-tetrafluorotoluene 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,3,5,6-Tetrafluorobenzylbromide2,3,5,6-Tetrafluorobenzyl bromide

L01916-06 L01916-06 L01916-03 L01916-03
AAAL01916-03EA 39.99 USD InStock
CAAAAL01916-03 CAAAAL01916-06 AAAL01916-03 AAAL01916-06
α-Bromo-2,3,5,6-tetrafluorotoluene 98+%
α-Bromo-2,3,5,6-tetrafluorotoluene


Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 79…80 °C/80 mm
Storage Temperature: Ambient
MDL Number: MFCD00013549
CAS Number: 53001-73-3
EINECS: 258-289-6
UN: 2922
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty