Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Thiophenemethanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Thiophen-3-ylmethanolThiophen-3-methanol3-Thienylmethanol

L01935-06 L01935-14 L01935-14 L01935-06
AAAL01935-14EA 375.99 USD InStock
CAAAAL01935-14 AAAL01935-14 CAAAAL01935-06 AAAL01935-06
3-Thiophenemethanol 98%
3-Thiophenemethanol


Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 86…88 °C/10 mm
Density: 1.210
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014534
CAS Number: 71637-34-8
EINECS: 275-741-8
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty