Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: 6-Amino-2-thiouracil hydrate

L01992-22 L01992-14 L01992-22 L01992-14
AAAL01992-14EA 27.79 USD InStock
AAAL01992-14 CAAAAL01992-14 CAAAAL01992-22 AAAL01992-22
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98%
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate
4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.


Formula: C₄H₅N₃OS·xH₂O (x=1)
MW: 143.17 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150556
CAS Number: 65802-56-4
EINECS: 213-722-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty