Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Ethyltoluene 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Ethyl-2-methylbenzene2-Ethyltoluene

L02028-06 L02028-06 L02028-14 L02028-14
AAAL02028-14EA 195.99 USD InStock
AAAL02028-14 CAAAAL02028-14 AAAL02028-06 CAAAAL02028-06
o-Ethyltoluene 98+%
o-Ethyltoluene


Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 164…165 °C
Melting Pt: –17 °C
Density: 0.884
Flash Pt: 53 °C(127 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009257
CAS Number: 611-14-3
EINECS: 210-255-1
UN: 3295
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty