Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxybenzylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: m-Anisylamine3-(Aminomethyl)anisole

L02054-14 L02054-14 L02054-06 L02054-06
AAAL02054-14EA 210.99 USD InStock
AAAL02054-14 CAAAAL02054-06 CAAAAL02054-14 AAAL02054-06
3-Methoxybenzylamine 98+%
3-Methoxybenzylamine


Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 230…231 °C
Density: 1.066
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008115
CAS Number: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty