Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Trichlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

L02132-18
AAAL02132-18EA 44.93 USD InStock
AAAL02132-18
1,3,5-Trichlorobenzene 98%
1,3,5-Trichlorobenzene


Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 208 °C
Melting Pt: 62…65 °C
Density: 1.4604 g/cm³ (20 °C)
Flash Pt: 126 °C(258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000585
CAS Number: 108-70-3
EINECS: 203-608-6
UN: 2811
ADR: "6.1",III
Merck Index: 12,09761
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty