Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isopentylamine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Isoamylamine3-Methylbutylamine1-Amino-3-methylbutaneLeucamineIsopentyl amine

L02260-14 L02260-14
AAAL02260-14EA 49.19 USD InStock
AAAL02260-14 CAAAAL02260-14
Isopentylamine 99%
Isopentylamine


Formula: C₅H₁₃N
MW: 87.16 g/mol
Boiling Pt: 95…97 °C
Melting Pt: –60 °C
Density: 0.751
Flash Pt: -1 °C(30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008203
CAS Number: 107-85-7
EINECS: 203-526-0
UN: 1106
ADR: "3",III
Merck Index: 13,05128
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty