Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydroxy-n-octanoic acid ≥98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Hydroxy-n-octanoic Acid

L02314-03 L02314-06 L02314-03 L02314-06
AAAL02314-03EA 38.13 USD InStock
CAAAAL02314-03 AAAL02314-03 CAAAAL02314-06 AAAL02314-06
2-Hydroxy-n-octanoic acid ≥98%
2-Hydroxy-n-octanoic acid


Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Melting Pt: 66…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00014410
CAS Number: 617-73-2
EINECS: 210-524-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty