Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2,2-dimethylpropane 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chloro-1-diméthyl-2,2-propaneneo-Pentylchloride

L02365-14 L02365-06 L02365-06 L02365-14
AAAL02365-14EA 207.99 USD InStock
CAAAAL02365-06 AAAL02365-14 CAAAAL02365-14 AAAL02365-06
1-Chloro-2,2-dimethylpropane 98%
1-Chloro-2,2-dimethylpropane


Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 83…84 °C
Melting Pt: –20 °C
Density: 0.868
Flash Pt: -8 °C(17 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000941
CAS Number: 753-89-9
EINECS: 212-040-8
UN: 1107
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty