Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzofuran-2-carboxaldehyde 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Benzo[b]furan-2-carboxaldehydeBenzofuran-2-carbaldehyde

L02456-09 L02456-04 L02456-09 L02456-04
AAAL02456-04EA 118 USD InStock
CAAAAL02456-04 AAAL02456-04 AAAL02456-09 CAAAAL02456-09
Benzofuran-2-carboxaldehyde 99%
Benzofuran-2-carboxaldehyde


Formula: C₉H₆O₂
MW: 146.145 g/mol
Boiling Pt: 92…97 °C/1 mm
Storage Temperature: Refrigerator
MDL Number: MFCD00015463
CAS Number: 4265-16-1
EINECS: 224-248-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty