Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylcyclohexylamine cis- and trans- mixture 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-(tert-Butyl)cyclohexanamine

L02472-14 L02472-06 L02472-06 L02472-14
AAAL02472-14EA 177.99 USD InStock
CAAAAL02472-14 CAAAAL02472-06 AAAL02472-14 AAAL02472-06
4-tert-Butylcyclohexylamine cis- and trans- mixture 97%
4-tert-Butylcyclohexylamine


Formula: C₁₀H₂₁N
MW: 155.28 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.870
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013763
CAS Number: 5400-88-4
EINECS: 226-444-7
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty