Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentyne 99%

Supplier: Thermo Scientific Chemicals
Warning

L02479-06 L02479-14 L02479-14 L02479-06
AAAL02479-14EA 98.9 USD InStock
CAAAAL02479-14 AAAL02479-14 CAAAAL02479-06 AAAL02479-06
1-Pentyne 99%
1-Pentyne


Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 39…41 °C
Melting Pt: –106 °C
Density: 0.692
Flash Pt: -20 °C(-4 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009469
CAS Number: 627-19-0
EINECS: 210-987-1
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty