Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hexynoic acid 96%

Supplier: Thermo Scientific Chemicals
Danger

L02526-06 L02526-06 L02526-03 L02526-03
AAAL02526-03EA 26.59 USD InStock
AAAL02526-03 AAAL02526-06 CAAAAL02526-06 CAAAAL02526-03
2-Hexynoic acid 96%
2-Hexynoic acid


Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 110…112 °C/10 mm
Melting Pt: 23…26 °C
Storage Temperature: Ambient
MDL Number: MFCD00014371
CAS Number: 764-33-0
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty