Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Tetradecyne tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals

L02573-06 L02573-14 L02573-14 L02573-06
AAAL02573-06EA 66.29 USD InStock
CAAAAL02573-14 AAAL02573-06 CAAAAL02573-06 AAAL02573-14
1-Tetradecyne tech. 90%, Technical Grade
1-Tetradecyne


Formula: C₁₄H₂₆
Boiling Pt: 132…134 °C/14 mm
Density: 0.790
Storage Temperature: Ambient
MDL Number: MFCD00015076
CAS Number: 765-10-6
EINECS: 212-139-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty