Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylvaleryl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

L02725-14 L02725-06 L02725-14 L02725-06
AAAL02725-06EA 39.05 USD InStock
AAAL02725-06 CAAAAL02725-06 AAAL02725-14 CAAAAL02725-14
2-Methylvaleryl chloride 98%
2-Methylvaleryl chloride


Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Boiling Pt: 140…144 °C
Density: 0.978
Flash Pt: 37 °C(98 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013656
CAS Number: 5238-27-7
EINECS: 226-035-3
UN: 2920
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty