Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexafluoroglutaric anhydride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3,3,4,4,5,5-Hexafluorodihydro-2H-pyran-2,6(3H)-dione

L02798-14 L02798-06 L02798-06 L02798-03 L02798-14 L02798-03
AAAL02798-06EA 86.49 USD InStock
AAAL02798-06 CAAAAL02798-06 AAAL02798-14 AAAL02798-03 CAAAAL02798-14 CAAAAL02798-03
Hexafluoroglutaric anhydride 97%
Hexafluoroglutaric anhydride


Formula: C₅F₆O₃
MW: 222.04 g/mol
Boiling Pt: 71…72 °C
Density: 1.652
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00006680
CAS Number: 376-68-1
EINECS: 206-811-8
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty