Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DL-1,2-IsopropylidenglycerolSolketalDL-1,2-Isopropylideneglycerol(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol1,2-O-Isopropylideneglycerol(±)-1,2-O-Isopropylidenglycerol2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

L02814-22 L02814-22 L02814-0E L02814-36 L02814-36 L02814-0E
AAAL02814-22EA 40.89 USD InStock
CAAAAL02814-22 AAAL02814-22 AAAL02814-0E CAAAAL02814-36 AAAL02814-36 CAAAAL02814-0E
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Useful for synthesis of mono-, di- and triglycerides


Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 188…190 °C
Melting Pt: –27 °C
Density: 1.064
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063238
CAS Number: 100-79-8
EINECS: 202-888-7
Merck Index: 13,05233
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty