Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Xylylenediamine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,4-Bis(aminomethyl)benzene1,4-Phenylenedimethanamine

L02995-09 L02995-04 L02995-04 L02995-18 L02995-18 L02995-09
AAAL02995-04EA 52.33 USD InStock
CAAAAL02995-04 CAAAAL02995-18 AAAL02995-04 AAAL02995-09 CAAAAL02995-09 AAAL02995-18
p-Xylylenediamine 97%
p-Xylylenediamine


Formula: C₈H₁₂N₂
MW: 136.20 g/mol
Boiling Pt: 230 °C/10 mm
Melting Pt: 59…63 °C
Flash Pt: 134 °C(273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009821
CAS Number: 539-48-0
EINECS: 208-719-3
UN: 3259
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty