Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butyronitrile 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: n-Butyronitrile

L02999-AK L02999-AU L02999-AU L02999-AK
AAAL02999-AUEA 84.19 USD InStock
AAAL02999-AU CAAAAL02999-AU CAAAAL02999-AK AAAL02999-AK
Butyronitrile 99%
Butyronitrile


Formula: C₄H₇N
MW: 69.11 g/mol
Boiling Pt: 115…117 °C
Melting Pt: –112 °C
Density: 0.792
Flash Pt: 18 °C(64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001968
CAS Number: 109-74-0
EINECS: 203-700-6
UN: 2411
ADR: "3",II
Merck Index: 13,01597
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty