Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-m-cresol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Hydroxy-3-methyl-4-nitrobenzene3-Methyl-4-nitrophenol

L03163-14 L03163-22 L03163-22 L03163-14
AAAL03163-14EA 46.21 USD InStock
AAAL03163-14 CAAAAL03163-14 AAAL03163-22 CAAAAL03163-22
4-Nitro-m-cresol 98%
4-Nitro-m-cresol


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00007333
CAS Number: 2581-34-2
EINECS: 219-952-5
UN: 2446
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty