Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thioacetic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

L03305-36 L03305-22 L03305-14 L03305-36 L03305-14 L03305-22
AAAL03305-36EA 150.99 USD InStock
CAAAAL03305-36 CAAAAL03305-14 AAAL03305-36 AAAL03305-14 AAAL03305-22 CAAAAL03305-22
Thioacetic acid 97%
Thioacetic acid


Formula: C₂H₄OS
MW: 76.12 g/mol
Boiling Pt: 97 °C
Melting Pt: –17 °C
Density: 1.068
Flash Pt: 18 °C(64 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004853
CAS Number: 507-09-5
EINECS: 208-063-8
UN: 2436
ADR: "3",II
Merck Index: 12,09454
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty