Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Aminouracil 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Amino-2,4-dihydroxypyrimidine4-Amino-2,6-dihydroxypyrimidine4-Aminouracil6-Aminopyrimidine-2,4(1H,3H)-dione

L03332-22 L03332-36 L03332-36 L03332-22
AAAL03332-36EA 192.99 USD InStock
CAAAAL03332-22 AAAL03332-36 CAAAAL03332-36 AAAL03332-22
6-Aminouracil 98%
6-Aminouracil


Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
Melting Pt: 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00006071
CAS Number: 873-83-6
EINECS: 212-854-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty