Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminophthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L03333-03 L03333-03 L03333-06 L03333-06
AAAL03333-06EA 166.99 USD InStock
AAAL03333-06 CAAAAL03333-06 AAAL03333-03 CAAAAL03333-03
4-Aminophthalic acid 98%
4-Aminophthalic acid


Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013985
CAS Number: 5434-21-9
EINECS: 226-596-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty