Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotributylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tributylchlorosilane

L03477-14 L03477-06 L03477-06 L03477-14
AAAL03477-06EA 64.59 USD InStock
AAAL03477-06 AAAL03477-14 CAAAAL03477-06 CAAAAL03477-14
Chlorotributylsilane 97%
Chlorotributylsilane


Formula: C₁₂H₂₇ClSi
MW: 234.88 g/mol
Boiling Pt: 93…94 °C/4 mm
Density: 0.883
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000509
CAS Number: 995-45-9
EINECS: 213-627-1
UN: 2987
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty