Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Lutidine 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3,4-Dimethylpyridine

L03489-AP L03489-AC L03489-AE L03489-AP L03489-AC L03489-AE
AAAL03489-APEA 226.99 USD InStock
AAAL03489-AP CAAAAL03489-AC CAAAAL03489-AP AAAL03489-AC CAAAAL03489-AE AAAL03489-AE
3,4-Lutidine 98+%
3,4-Lutidine


Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 163…164 °C
Melting Pt: –11 °C
Density: 0.955
Flash Pt: 47 °C(116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006403
CAS Number: 583-58-4
EINECS: 209-511-5
UN: 2929
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty