Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl sulfide 96%

Supplier: Thermo Scientific Chemicals
Danger

L03493-22 L03493-09 L03493-09 L03493-22
AAAL03493-09EA 47.25 USD InStock
CAAAAL03493-09 AAAL03493-09 CAAAAL03493-22 AAAL03493-22
Diethyl sulfide 96%
Diethyl sulphide


Formula: C₄H₁₀S
MW: 90.19 g/mol
Boiling Pt: 90…92 °C
Melting Pt: –104 °C
Density: 0.835
Flash Pt: -10 °C(14 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009270
CAS Number: 352-93-2
EINECS: 206-526-9
UN: 2375
ADR: "3",II
Merck Index: 13,03885
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty