Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Lutidine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,3-Dimethylpyridine

L03697-14 L03697-22 L03697-36 L03697-14 L03697-36 L03697-22
AAAL03697-14EA 34.49 USD InStock
AAAL03697-14 AAAL03697-36 CAAAAL03697-14 CAAAAL03697-36 CAAAAL03697-22 AAAL03697-22
2,3-Lutidine 99%
2,3-Lutidine


Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 160…161 °C
Melting Pt: –17 °C
Density: 0.948
Flash Pt: 50 °C(122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009605
CAS Number: 583-61-9
EINECS: 209-514-1
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty