Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Naphthonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Naphthalene-2-carbonitrile

L03704-14 L03704-14
AAAL03704-14EA 227.99 USD InStock
CAAAAL03704-14 AAAL03704-14
2-Naphthonitrile 97%
2-Naphthonitrile


Formula: C₁₁H₇N
MW: 153.18 g/mol
Boiling Pt: 156…158 °C/12 mm
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00016807
CAS Number: 613-46-7
EINECS: 210-344-5
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty