Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-chlorobutyrate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl 4-chlorobutyrate

L03734-22 L03734-14 L03734-14 L03734-22
AAAL03734-22EA 70.99 USD InStock
AAAL03734-22 CAAAAL03734-22 AAAL03734-14 CAAAAL03734-14
Methyl-4-chlorobutyrate 98%
Methyl 4-chlorobutyrate


Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 175…176 °C
Density: 1.125
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001003
CAS Number: 3153-37-5
EINECS: 221-592-9
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty