Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl Phenylsulfonylacetate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Phenylsulphonylacetic Acid Methyl EsterPhenylsulfonylacetic Acid Methyl Ester

L03748-14 L03748-06 L03748-14 L03748-06
AAAL03748-14EA 85.79 USD InStock
AAAL03748-14 CAAAAL03748-14 AAAL03748-06 CAAAAL03748-06
Methyl Phenylsulfonylacetate 98%
Methyl Phenylsulfonylacetate


Formula: C₉H₁₀O₄S
MW: 214.24 g/mol
Boiling Pt: 165 °C/0.05 mm
Melting Pt: ca 30 °C
Density: 1.240
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007555
CAS Number: 34097-60-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty