Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Methyl-8-nitroquinoline 98+%

Supplier: Thermo Scientific Chemicals
Warning

L03882-14 L03882-14 L03882-06 L03882-06
AAAL03882-14EA 409.99 USD InStock
CAAAAL03882-06 AAAL03882-14 AAAL03882-06 CAAAAL03882-14
7-Methyl-8-nitroquinoline 98+%
7-Methyl-8-nitroquinoline


Formula: C₁₀H₈N₂O₂
Melting Pt: 185…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00041234
CAS Number: 7471-63-8
EINECS: 231-268-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty