Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isobutyric acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methylpropionic acid2-Methylpropanoic acid

L04038-AE L04038-AE L04038-0F L04038-AP L04038-0F L04038-AP
AAAL04038-APEA 49.84 USD InStock
CAAAAL04038-AP AAAL04038-AP CAAAAL04038-AE CAAAAL04038-0F AAAL04038-AE AAAL04038-0F
Isobutyric acid 99%
Isobutyric acid


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 153…154 °C
Melting Pt: –47 °C
Density: 0.948
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002658
CAS Number: 79-31-2
EINECS: 201-195-7
UN: 2529
ADR: "3",III
Merck Index: 13,05172
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty