Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-N-trimethylsilylacetamide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Methyl-N-TMS-acetamide

L04101-09 L04101-09 L04101-04 L04101-04
AAAL04101-04EA 34.39 USD InStock
CAAAAL04101-04 AAAL04101-04 CAAAAL04101-09 AAAL04101-09
N-Methyl-N-trimethylsilylacetamide 98%
N-Methyl-N-trimethylsilylacetamide


Formula: C₆H₁₅NOSi
MW: 145.28 g/mol
Boiling Pt: 48…49 °C/11 mm
Density: 0.906
Flash Pt: 48 °C(118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040758
CAS Number: 7449-74-3
EINECS: 231-217-0
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty