Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Ethylthio)acetone 95%

Supplier: Thermo Scientific Chemicals
Warning

L04142-14 L04142-06 L04142-14 L04142-06
AAAL04142-14EA 152.99 USD InStock
CAAAAL04142-06 AAAL04142-14 AAAL04142-06 CAAAAL04142-14
(Ethylthio)acetone 95%
(Ethylthio)acetone


Formula: C₅H₁₀OS
Boiling Pt: 170…172 °C
Flash Pt: 52 °C(125 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026976
CAS Number: 20996-62-7
EINECS: 244-143-9
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty