Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citraconic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methyl-2-butenedioic acid

L04178-09 L04178-09 L04178-18 L04178-18
AAAL04178-09EA 35.49 USD InStock
CAAAAL04178-18 AAAL04178-09 CAAAAL04178-09 AAAL04178-18
Citraconic acid 98+%
Citraconic acid


Formula: C₅H₆O₄
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 88…91 °C
Density: 1.620
Storage Temperature: Ambient
MDL Number: MFCD00078086
CAS Number: 498-23-7
EINECS: 207-858-7
Merck Index: 13,02345
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty