Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propionic acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propanoic acid

L04210-0E L04210-0E L04210-22 L04210-0B L04210-22 L04210-0B
AAAL04210-22EA 46.53 USD InStock
CAAAAL04210-0B AAAL04210-22 CAAAAL04210-22 CAAAAL04210-0E AAAL04210-0B AAAL04210-0E
Propionic acid 99%
Propionic acid


Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 140…141 °C
Melting Pt: –21 °C
Density: 0.992
Flash Pt: 51 °C(123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002756
CAS Number: 79-09-4
EINECS: 201-176-3
UN: 3463
ADR: "8",II
REACH: 01-2119486971-24
Merck Index: 13,07917
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty