Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloro(phenyl)silane 96%

Supplier: Thermo Scientific Chemicals
Danger

L04371-06 L04371-14 L04371-14 L04371-06
AAAL04371-06EA 54.19 USD InStock
AAAL04371-06 CAAAAL04371-14 CAAAAL04371-06 AAAL04371-14
Dichloro(phenyl)silane 96%
Dichloro(phenyl)silane


Formula: C₆H₆Cl₂Si
Boiling Pt: 181 °C
Density: 1.212
Flash Pt: 48 °C(118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039296
CAS Number: 1631-84-1
EINECS: 216-635-3
UN: 2986
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty