Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(3-Hydroxypropyl)phthalimide 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-(3-Hydroxypropyl)isoindoline-1,3-dione

L04507-09 L04507-09 L04507-18 L04507-18
AAAL04507-09EA 28.49 USD InStock
AAAL04507-09 CAAAAL04507-09 AAAL04507-18 CAAAAL04507-18
N-(3-Hydroxypropyl)phthalimide 98%
N-(3-Hydroxypropyl)phthalimide


Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
Melting Pt: 75…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00023097
CAS Number: 883-44-3
EINECS: 212-931-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty