Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-1,3-dinitrobenzene 98+%

Supplier: Thermo Scientific Chemicals
Danger

L04593-06 L04593-03 L04593-03 L04593-06
AAAL04593-06EA 220.99 USD InStock
CAAAAL04593-03 AAAL04593-06 CAAAAL04593-06 AAAL04593-03
2-Chloro-1,3-dinitrobenzene 98+%
2-Chloro-1,3-dinitrobenzene


Formula: C₆H₃ClN₂O₄
Boiling Pt: 315 °C
Melting Pt: 83…88 °C
Density: 1.687
Storage Temperature: Ambient
MDL Number: MFCD00014689
CAS Number: 606-21-3
EINECS: 210-107-6
UN: 3441
ADR: "6.1",II
Merck Index: 13,02156
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty