Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-1-Phenyl-1-pentanol 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Fenipentol

L04646-09 L04646-09 L04646-18 L04646-18
AAAL04646-18EA 133.99 USD InStock
CAAAAL04646-09 AAAL04646-18 CAAAAL04646-18 AAAL04646-09
(±)-1-Phenyl-1-pentanol 95%
(±)-1-Phenyl-1-pentanol


Formula: C₆H₅CH(OH)(CH₂)₃CH₃
MW: 164.25 g/mol
Boiling Pt: 137 °C/2 mm
Density: 0.960
Storage Temperature: Ambient
MDL Number: MFCD00021935
CAS Number: 583-03-9
EINECS: 209-493-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty