Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Butyl phenyl ether 99%

Supplier: Thermo Scientific Chemicals
Warning

L04703-14 L04703-14 L04703-22 L04703-22
AAAL04703-14EA 38.69 USD InStock
AAAL04703-14 AAAL04703-22 CAAAAL04703-14 CAAAAL04703-22
n-Butyl phenyl ether 99%
n-Butyl phenyl ether


Formula: C₁₀H₁₄O
Boiling Pt: 210…211 °C
Melting Pt: –19 °C
Density: 0,938
Flash Pt: 82 °C(179 °F)
Storage Temperature: Ambient
EINECS: 214-426-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty