Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(3-Aminopropyl)pyrrolidine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-(Pyrrolidin-1-yl)propan-1-amineN-(3-Aminopropyl)pyrrolidine

L04739-06 L04739-14 L04739-14 L04739-06
AAAL04739-14EA 260.99 USD InStock
CAAAAL04739-06 AAAL04739-14 CAAAAL04739-14 AAAL04739-06
1-(3-Aminopropyl)pyrrolidine 97%
1-(3-Aminopropyl)pyrrolidine


Formula: C₇H₁₆N₂
MW: 128.22 g/mol
Boiling Pt: 191…193 °C
Density: 1.014
Storage Temperature: Ambient
MDL Number: MFCD00014100
CAS Number: 23159-07-1
EINECS: 245-465-2
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty