Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromoheptane 97%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

L04807-06 L04807-14 L04807-06 L04807-14
AAAL04807-14EA 176.52 USD InStock
CAAAAL04807-14 CAAAAL04807-06 AAAL04807-14 AAAL04807-06
4-Bromoheptane 97%, Technical Grade
4-Bromoheptane


Formula: H₃C(CH₂)₂BrCH(CH₂)₂CH₃
MW: 179.1 g/mol
Boiling Pt: 60…62 °C/18 mm
Density: 1.140
Flash Pt: 40 °C(104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039182
CAS Number: 998-93-6
EINECS: 213-653-3
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty