Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dibromobutyric acid 97%

Supplier: Thermo Scientific Chemicals
Danger

L04841-06 L04841-14 L04841-06 L04841-14
AAAL04841-14EA 82.69 USD InStock
CAAAAL04841-06 AAAL04841-14 CAAAAL04841-14 AAAL04841-06
2,3-Dibromobutyric acid 97%
2,3-Dibromobutyric acid


Formula: H₃C(BrCH)₂COOH
Boiling Pt: 100…110 °C/2 mm
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00014334
CAS Number: 600-30-6
EINECS: 209-992-1
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty