Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Phenylenanthic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 7-Phenylheptanoic acid

L05009-06 L05009-03 L05009-06 L05009-03
AAAL05009-06EA 378.99 USD InStock
CAAAAL05009-06 AAAL05009-06 CAAAAL05009-03 AAAL05009-03
7-Phenylenanthic acid 97%
7-Phenylenanthic acid


Formula: C₁₃H₁₈O₂
MW: 206.29 g/mol
Boiling Pt: 147…150 °C/0.4 mm
Melting Pt: ca 20 °C
Density: 1,009
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037801
CAS Number: 40228-90-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty