Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl cholate 98+%

Supplier: Thermo Scientific Chemicals

L05129-06 L05129-14 L05129-06 L05129-14
AAAL05129-06EA 48.83 USD InStock
AAAL05129-06 AAAL05129-14 CAAAAL05129-06 CAAAAL05129-14
Methyl cholate 98+%
Methyl cholate


Formula: C₂₅H₄₂O₅
MW: 422.61 g/mol
Melting Pt: 155…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00064934
CAS Number: 1448-36-8
EINECS: 215-903-7
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty